Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700013742.mm.1 |
TC0700001651.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700013742.mm.1 |
NM_001166584.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700013742.mm.1 |
NM_001166584.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700013742.mm.1 |
NM_001166585.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700013742.mm.1 |
NM_001166585.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700013742.mm.1 |
NM_009346.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700013742.mm.1 |
NM_009346.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700013744.mm.1 |
TC0700001651.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700013744.mm.1 |
NM_001166584.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700013744.mm.1 |
NM_001166584.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700013744.mm.1 |
NM_001166585.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700013744.mm.1 |
NM_001166585.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700013744.mm.1 |
NM_009346.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700013744.mm.1 |
NM_009346.3 |
Refseq |