Assay Details
| Gene Symbol: Far1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285831.1 | NP_001272760.1 | 2 - 3 | 155 | 67 | |
| NM_027379.3 | NP_081655.2 | 2 - 3 | 155 | 67 | ||
| GenBank mRNA | AK005531.1 | - | 2 - 3 | 129 | 67 | |
| AK011187.1 | - | 2 - 3 | 130 | 67 | ||
| AK014405.1 | - | 2 - 3 | 128 | 67 | ||
| AK014486.1 | - | 2 - 3 | 128 | 67 | ||
| AK030067.1 | - | 2 - 3 | 86 | 67 | ||
| AK033674.1 | - | 2 - 3 | 98 | 67 | ||
| BC007178.1 | - | 2 - 3 | 103 | 3495305 | 67 | |
| BY323527.1 | - | 2 - 3 | 219 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005531.1, AK011187.1, AK014486.1, AK030067.1, AK033674.1, BC007178.1, NM_001285831.1, NM_027379.3 transcripts.
Set Membership: |
|

Far1
Genomic Map