Assay Details
| Gene Symbol: Far1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285831.1 | NP_001272760.1 | 8 - 9 | 1050 | 66 | |
| NM_027379.3 | NP_081655.2 | 8 - 9 | 1050 | 66 | ||
| GenBank mRNA | AK005531.1 | - | 8 - 9 | 1024 | 66 | |
| AK014405.1 | - | 8 - 9 | 1023 | 66 | ||
| AK014486.1 | - | 8 - 9 | 1023 | 66 | ||
| AK030067.1 | - | 8 - 9 | 981 | 66 | ||
| AK033674.1 | - | 8 - 9 | 993 | 66 | ||
| AK039083.1 | - | 4 - 5 | 468 | 66 | ||
| BC007178.1 | - | 8 - 9 | 998 | 3495305 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Far1
Genomic Map