Assay Details
| Gene Symbol: Slc23a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018824.2 | NP_061294.2 | 12 - 13 | 1591 | 81 | |
| GenBank mRNA | AB038145.1 | - | 11 - 12 | 1327 | 81 | |
| AK122227.1 | - | 8 - 9 | 1363 | 81 | ||
| AY004874.1 | - | 12 - 13 | 1545 | 81 | ||
| BC050823.1 | - | 11 - 12 | 1355 | 6332451 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc23a2
Genomic Map