Assay Details
| Gene Symbol: Slc23a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018824.2 | NP_061294.2 | 3 - 4 | 449 | 63 | |
| GenBank mRNA | AB038145.1 | - | 2 - 3 | 185 | 63 | |
| AK087175.1 | - | 3 - 4 | 457 | 63 | ||
| AK169991.1 | - | 3 - 4 | 451 | 63 | ||
| AY004874.1 | - | 3 - 4 | 403 | 63 | ||
| BC050823.1 | - | 2 - 3 | 213 | 6332451 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc23a2
Genomic Map