Assay Details
| Gene Symbol: Slc23a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018824.2 | NP_061294.2 | 8 - 9 | 1062 | 132 | |
| GenBank mRNA | AB038145.1 | - | 7 - 8 | 798 | 132 | |
| AK122227.1 | - | 4 - 5 | 908 | 132 | ||
| AY004874.1 | - | 8 - 9 | 1016 | 132 | ||
| BC050823.1 | - | 7 - 8 | 826 | 6332451 | 132 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc23a2
Genomic Map