Assay Details
| Gene Symbol: Pde3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011055.2 | NP_035185.2 | 10 - 11 | 2428 | 166 | |
| GenBank mRNA | AF547435.1 | - | 10 - 11 | 2401 | 166 | |
| AJ132271.1 | - | 10 - 11 | 2131 | 166 | ||
| BU613166.1 | - | 2 - 3 | 137 | 166 | ||
| X95521.1 | - | 10 - 11 | 1273 | 166 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pde3b
Genomic Map