Assay Details
| Gene Symbol: Pde3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011055.2 | NP_035185.2 | 3 - 4 | 1508 | 92 | |
| GenBank mRNA | AF547435.1 | - | 3 - 4 | 1481 | 92 | |
| AJ132271.1 | - | 3 - 4 | 1211 | 92 | ||
| BY745829.1 | - | 3 - 4 | 411 | 92 | ||
| X95521.1 | - | 3 - 4 | 353 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pde3b
Genomic Map