Assay Details
| Gene Symbol: Pde3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011055.2 | NP_035185.2 | 4 - 5 | 1648 | 110 | |
| GenBank mRNA | AF547435.1 | - | 4 - 5 | 1621 | 110 | |
| BY745829.1 | - | 4 - 5 | 110 | |||
| X95521.1 | - | 4 - 5 | 493 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pde3b
Genomic Map