Assay Details
| Gene Symbol: Pacsin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289677.1 | NP_001276606.1 | 7 - 8 | 1113 | 85 | |
| NM_001289678.1 | NP_001276607.1 | 8 - 9 | 1112 | 85 | ||
| NM_028733.4 | NP_083009.1 | 8 - 9 | 1242 | 85 | ||
| NM_030880.3 | NP_112019.2 | 8 - 9 | 1108 | 85 | ||
| GenBank mRNA | AF149824.1 | - | 8 - 9 | 1026 | 85 | |
| AF242531.1 | - | 7 - 8 | 1011 | 85 | ||
| AK049166.1 | - | 8 - 9 | 1243 | 85 | ||
| AK078060.1 | - | 7 - 8 | 1119 | 85 | ||
| AK141063.1 | - | 8 - 9 | 1085 | 85 | ||
| BC003884.1 | - | 8 - 9 | 1049 | 3497435 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pacsin3
Genomic Map