Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200037851.mm.1 |
NM_013735.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037851.mm.1 |
TC0200004482.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037850.mm.1 |
NM_013735.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037850.mm.1 |
TC0200004482.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037852.mm.1 |
NM_013735.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037852.mm.1 |
TC0200004482.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037848.mm.1 |
NM_013735.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037848.mm.1 |
TC0200004482.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037849.mm.1 |
NM_013735.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037849.mm.1 |
TC0200004482.mm.1 |
MTA TC |