Assay Details
Gene Symbol: Trp53bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290830.1 | NP_001277759.1 | 3 - 4 | 413 | 103 | |
NM_013735.4 | NP_038763.3 | 3 - 4 | 413 | 103 | ||
GenBank mRNA | AJ414734.1 | - | 2 - 3 | 277 | 103 | |
AK045851.1 | - | 3 - 4 | 328 | 103 | ||
AK148123.1 | - | 3 - 4 | 376 | 103 | ||
BC037378.1 | - | 3 - 4 | 375 | 103 | ||
BC079906.1 | - | 3 - 4 | 347 | 30639375 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
