Assay Details
| Gene Symbol: Pax6os1 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002867.2 | 3 - 4 | 305 | 134 | |||
| GenBank mRNA | AK029183.1 | - | LIT3400 Pax6OS |
3 - 4 | 306 | 134 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | reticulocalbin 1 | Rcn1 | Rcn | -136.8kb | Sense | Chr. 2 - 105385948 - 105399319 on NCBI Build GRCm38 |
| Downstream | Pax6 upstream antisense RNA | Paupar | 72.9kb | Sense | Chr. 2 - 105657862 - 105661343 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P122160 | NR_002867.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P122160 | AK029183.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 157005 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 589828 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 330218 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 721144 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 442398 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 682399 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 484454 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 28997 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 763550 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 893478 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 420895 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 520771 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 807631 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 626414 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 881259 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 331717 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 238628 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 209130 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 545392 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 898510 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 402809 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 118306 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 172226 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 725418 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 438069 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 595691 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 613281 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 648686 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 309123 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 752975 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 971718 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 136599 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61921 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 16857 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 852858 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 327872 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916199 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 833289 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 749388 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 908895 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 125198 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 153847 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 461442 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 679908 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 409844 | AK043788.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 351092 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 887934 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 137415 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782489 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 850596 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 440477 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 409526 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 651019 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 158823 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 223110 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 591080 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 421757 | AK045712.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 41009 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 497758 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 372013 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 45431 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 9616 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 89591 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 207231 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 625539 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 425519 | AJ245945.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 425519 | AK140173.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 425519 | AK165775.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 425519 | BC024263.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 425519 | NM_022414.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 629800 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930257 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814503 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 237407 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 629987 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 764687 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 945729 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 59543 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 301033 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 472620 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 458645 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 213979 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 570825 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 92583 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 41867 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 21405 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 140913 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 125025 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 177563 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 146680 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 94681 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 34999 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 146868 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 68425 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 22667 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103939 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116827 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P122160 | NR_002867.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P122160 | AK029183.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00452087 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04634770 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04634823 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04634707 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04634801 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00451976 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116827 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103939 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04634639 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122160 | NR_002867.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122160 | AK029183.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103939 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116827 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00452087 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04634801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01843158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495596 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01504082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495209 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01195232 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495579 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01843155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01812410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495730 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495574 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495527 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495600 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01659472 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495531 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495593 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495727 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01843163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495592 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01176648 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495753 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495729 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02209631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495656 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01255001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495567 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01195230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495568 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495540 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495697 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495575 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495667 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01281816 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01281819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495720 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495571 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495732 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495648 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02004679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01935838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495728 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02168337 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01812403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495598 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495583 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495674 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495526 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495576 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02168333 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495654 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495566 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495811 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495692 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495584 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495693 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495528 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495577 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01935841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495808 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01458718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01812418 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495213 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495813 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495715 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02209630 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495711 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01452445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01874831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01168391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01417847 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21495589 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1440445_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440445_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1440445_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 402728_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 402728_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200035882.mm.1 | NR_002867.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200035882.mm.1 | NR_002867.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200035882.mm.1 | TC0200004265.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200035880.mm.1 | NR_002867.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200035880.mm.1 | NR_002867.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200035880.mm.1 | TC0200004265.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200035881.mm.1 | NR_002867.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200035881.mm.1 | NR_002867.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200035881.mm.1 | TC0200004265.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1440445_PM_at | - | - |
| Mouse Expression Set 430 | 1440445_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440445_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440445_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 752409 | - | - |

Pax6os1
Genomic Map