Assay Details
| Gene Symbol: Hbb-y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008221.4 | NP_032247.1 | 2 - 3 | 371 | 67 | |
| GenBank mRNA | AK011619.1 | - | 2 - 3 | 372 | 67 | |
| AK012501.1 | - | 2 - 3 | 373 | 67 | ||
| AK012897.1 | - | 2 - 3 | 372 | 67 | ||
| BC057014.1 | - | 2 - 3 | 348 | 6817810 | 67 | |
| J00415.1 | - | 1 - 2 | 79 | 67 | ||
| M26897.1 | - | 2 - 3 | 333 | 67 | ||
| V00857.1 | - | 2 - 3 | 252 | 67 | ||
More Information
Set Membership: |
|

Hbb-y
Genomic Map