Assay Details
| Gene Symbol: Olfr700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146600.1 | NP_666811.1 | 1 | 44 | 72 | |
| GenBank mRNA | BC104090.1 | - | 1 | 44 | 72 | |
| BC104091.2 | - | 1 | 44 | 40042950 | 72 | |
| BC106811.1 | - | 1 | 44 | 72 | ||
| BC106812.2 | - | 1 | 44 | 40043224 | 72 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Olfr700
Genomic Map