Assay Details
| Gene Symbol: Gzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028986.3 | NP_083262.1 | 2 - 3 | 1473 | 79 | |
| GenBank mRNA | AB100266.1 | - | 2 - 3 | 1402 | 79 | |
| AK018470.1 | - | 2 - 3 | 1403 | 79 | ||
| AK052820.1 | - | 2 - 3 | 1430 | 79 | ||
| AK053262.1 | - | 2 - 3 | 1430 | 79 | ||
| AK141153.1 | - | 2 - 3 | 1449 | 79 | ||
| BC096015.1 | - | 2 - 3 | 1395 | 6821149 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gzf1
Genomic Map