Assay Details
Gene Symbol: Prdm11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177536.1 | NP_001171007.1 | 3 - 4 | 395 | 72 | |
GenBank mRNA | AK078754.1 | - | 2 - 3 | 250 | 72 | |
BE648497.1 | - | 3 - 4 | 72 | |||
CJ069425.1 | - | 2 - 3 | 243 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
methylation
rRNA (adenine-N6,N6-)-dimethyltransferase activity
molecular_function
protein C-terminal carboxyl O-methyltransferase activity
tRNA (guanine-N2-)-methyltransferase activity
methyltransferase activity
C-methyltransferase activity
N-methyltransferase activity
O-methyltransferase activity
S-methyltransferase activity
RNA methyltransferase activity
mRNA methyltransferase activity
tRNA methyltransferase activity
protein methyltransferase activity
2-polyprenyl-6-methoxy-1,4-benzoquinone methyltransferase activity
rRNA methyltransferase activity
rRNA (uridine-2'-O-)-methyltransferase activity
rRNA (adenine-N6-)-methyltransferase activity
DNA-methyltransferase activity
tRNA (guanine-N1-)-methyltransferase activity
rRNA (cytosine-C5-)-methyltransferase activity
selenocysteine methyltransferase activity
arginine N-methyltransferase activity
protein-arginine N-methyltransferase activity
lysine N-methyltransferase activity
protein-lysine N-methyltransferase activity
tRNA (uracil) methyltransferase activity
tRNA (guanine) methyltransferase activity
tRNA (adenine) methyltransferase activity
tRNA (cytosine) methyltransferase activity
tRNA (cytosine-5-)-methyltransferase activity
tRNA (adenine-N1-)-methyltransferase activity
rRNA (adenine) methyltransferase activity
rRNA (cytosine) methyltransferase activity
rRNA (guanine) methyltransferase activity
rRNA (uridine) methyltransferase activity
transferase activity
protein C-terminal leucine carboxyl O-methyltransferase activity
1-phenanthrol methyltransferase activity
protein-arginine N5-methyltransferase activity
methylarsonite methyltransferase activity
dimethylarsinite methyltransferase activity
4,5-dihydroxybenzo(a)pyrene methyltransferase activity
1-hydroxypyrene methyltransferase activity
1-hydroxy-6-methoxypyrene methyltransferase activity
demethylmenaquinone methyltransferase activity
cobalt-precorrin-6B C5-methyltransferase activity
cobalt-precorrin-7 C15-methyltransferase activity
cobalt-precorrin-5B C1-methyltransferase activity
cobalt-precorrin-3 C17-methyltransferase activity
dimethylamine methyltransferase activity
hydroxyneurosporene-O-methyltransferase activity
tRNA (adenine-57, 58-N(1)-) methyltransferase activity
methylamine-specific methylcobalamin:coenzyme M methyltransferase activity
trimethylamine methyltransferase activity
methanol-specific methylcobalamin:coenzyme M methyltransferase activity
monomethylamine methyltransferase activity
P-methyltransferase activity
Se-methyltransferase activity
2-phytyl-1,4-naphthoquinone methyltransferase activity
tRNA (uracil-2'-O-)-methyltransferase activity
tRNA (cytosine-2'-O-)-methyltransferase activity
phosphomethylethanolamine N-methyltransferase activity
tRNA (cytosine-3-)-methyltransferase activity
rRNA (cytosine-2'-O-)-methyltransferase activity
rRNA (cytosine-N4-)-methyltransferase activity
trihydroxyferuloyl spermidine O-methyltransferase activity
biological_process
methylation
rRNA (adenine-N6,N6-)-dimethyltransferase activity
molecular_function
protein C-terminal carboxyl O-methyltransferase activity
tRNA (guanine-N2-)-methyltransferase activity
methyltransferase activity
C-methyltransferase activity
N-methyltransferase activity
O-methyltransferase activity
S-methyltransferase activity
RNA methyltransferase activity
mRNA methyltransferase activity
tRNA methyltransferase activity
protein methyltransferase activity
2-polyprenyl-6-methoxy-1,4-benzoquinone methyltransferase activity
rRNA methyltransferase activity
rRNA (uridine-2'-O-)-methyltransferase activity
rRNA (adenine-N6-)-methyltransferase activity
DNA-methyltransferase activity
tRNA (guanine-N1-)-methyltransferase activity
rRNA (cytosine-C5-)-methyltransferase activity
selenocysteine methyltransferase activity
arginine N-methyltransferase activity
protein-arginine N-methyltransferase activity
lysine N-methyltransferase activity
protein-lysine N-methyltransferase activity
tRNA (uracil) methyltransferase activity
tRNA (guanine) methyltransferase activity
tRNA (adenine) methyltransferase activity
tRNA (cytosine) methyltransferase activity
tRNA (cytosine-5-)-methyltransferase activity
tRNA (adenine-N1-)-methyltransferase activity
rRNA (adenine) methyltransferase activity
rRNA (cytosine) methyltransferase activity
rRNA (guanine) methyltransferase activity
rRNA (uridine) methyltransferase activity
transferase activity
protein C-terminal leucine carboxyl O-methyltransferase activity
1-phenanthrol methyltransferase activity
protein-arginine N5-methyltransferase activity
methylarsonite methyltransferase activity
dimethylarsinite methyltransferase activity
4,5-dihydroxybenzo(a)pyrene methyltransferase activity
1-hydroxypyrene methyltransferase activity
1-hydroxy-6-methoxypyrene methyltransferase activity
demethylmenaquinone methyltransferase activity
cobalt-precorrin-6B C5-methyltransferase activity
cobalt-precorrin-7 C15-methyltransferase activity
cobalt-precorrin-5B C1-methyltransferase activity
cobalt-precorrin-3 C17-methyltransferase activity
dimethylamine methyltransferase activity
hydroxyneurosporene-O-methyltransferase activity
tRNA (adenine-57, 58-N(1)-) methyltransferase activity
methylamine-specific methylcobalamin:coenzyme M methyltransferase activity
trimethylamine methyltransferase activity
methanol-specific methylcobalamin:coenzyme M methyltransferase activity
monomethylamine methyltransferase activity
P-methyltransferase activity
Se-methyltransferase activity
2-phytyl-1,4-naphthoquinone methyltransferase activity
tRNA (uracil-2'-O-)-methyltransferase activity
tRNA (cytosine-2'-O-)-methyltransferase activity
phosphomethylethanolamine N-methyltransferase activity
tRNA (cytosine-3-)-methyltransferase activity
rRNA (cytosine-2'-O-)-methyltransferase activity
rRNA (cytosine-N4-)-methyltransferase activity
trihydroxyferuloyl spermidine O-methyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2130565 | NM_001177536.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P167117 | AK078754.1 | mouse_transcriptome |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21432015 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01531628 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21431977 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21432007 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21431958 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21432014 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21431955 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21431951 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21431968 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21431974 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21431986 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21432006 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21432011 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21431984 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21431952 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21431965 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01149753 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01851963 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21431998 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21431975 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02100391 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21431954 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21431971 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21432001 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21431948 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02100398 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21432010 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02100376 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21432016 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21431979 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02112126 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21431957 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21431967 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21432024 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21432021 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21431972 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02100403 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21432004 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02100375 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21432019 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21431999 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21431989 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21432009 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02100361 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21432020 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21431981 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10485197 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10485196 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10485195 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1436651_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1456186_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10485191 | - | - |
Mouse Genome 430 2.0 Array | 1436651_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1456186_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1456186_at | - | - |
Mouse Genome 430 2.0 Array | 1436651_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0200034829.mm.1 | NM_001177536.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200034829.mm.1 | NM_001177536.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200034829.mm.1 | TC0200004148.mm.1 | MTA TC |
Mouse Expression Set 430 | 1436651_PM_at | - | - |
Mouse Expression Set 430 | 1456186_at | - | - |
Mouse Expression Set 430 | 1456186_PM_at | - | - |
Mouse Expression Set 430 | 1436651_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1436651_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456186_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1436651_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456186_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 327248 | - | - |
