Assay Details
| Gene Symbol: Tspan18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183180.2 | NP_899003.1 | 7 - 8 | 824 | 77 | |
| GenBank mRNA | AK032765.1 | - | 6 - 7 | 735 | 77 | |
| BC052169.1 | - | 7 - 8 | 787 | 6335221 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tspan18
Genomic Map