Assay Details
| Gene Symbol: 4930405A10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046307.1 | 2 - 3 | 195 | 92 | ||
| GenBank mRNA | AK015475.1 | - | 2 - 3 | 197 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 503 | Zfp503 |
AI181838, B830002A16Rik, Nolz-1, Nolz1, Znf503 |
-541.4kb | Sense | Chr. 14 - 21983962 - 21989601 on NCBI Build GRCm38 |
| Downstream | predicted gene 10248 | Gm10248 | ENSMUSG00000068680 | 503.5kb | Sense | Chr. 14 - 23038194 - 23094571 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930405A10Rik
Genomic Map