Assay Details
| Gene Symbol: 4930405A10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046307.1 | 3 - 4 | 383 | 92 | ||
| GenBank mRNA | AK015475.1 | - | 3 - 4 | 385 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 503 | Zfp503 |
AI181838, B830002A16Rik, Nolz-1, Nolz1, Znf503 |
-517.6kb | Sense | Chr. 14 - 21983962 - 21989601 on NCBI Build GRCm38 |
| Downstream | predicted gene 10248 | Gm10248 | ENSMUSG00000068680 | 507.0kb | Sense | Chr. 14 - 23038194 - 23094571 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2248150 | AK015475.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P179909 | AK015091.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P179909 | AK015475.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114992 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11132731 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11166091 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459516 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459476 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11162503 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459364 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459354 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459497 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11162511 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459350 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11238135 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11206704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459469 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459479 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459428 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459459 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11713689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459593 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11501626 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459618 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459489 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459586 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459621 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459464 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459317 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11733040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11288453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459567 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459381 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11640000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459588 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459361 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459541 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459356 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459511 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11330275 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459387 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459400 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459404 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459490 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11534035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459499 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459612 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11238133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459501 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459520 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11534036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459324 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459460 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459564 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459369 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11367024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459536 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459472 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459617 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459370 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459531 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459551 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11730823 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11132730 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459526 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459527 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459335 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459592 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459560 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459595 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459512 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459475 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459431 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459430 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11288456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459570 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459572 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459441 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459343 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459614 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459542 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11330274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11626453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459510 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459371 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459435 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459532 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459342 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459513 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11363238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11361057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459448 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459461 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459402 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459447 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459529 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459336 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459449 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459384 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459351 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11640002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459454 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11626450 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459386 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459473 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459519 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459355 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459439 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459344 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11639988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459328 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459543 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459504 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459426 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459405 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459316 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459394 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459494 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11367029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11501639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459482 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459388 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11688255 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459478 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459352 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459581 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11162506 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459576 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459397 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459492 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459348 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11249338 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459450 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459389 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459508 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11577558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11623120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11414597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459340 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459391 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459458 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459601 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459338 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459483 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459535 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459578 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11537118 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459506 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459603 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459615 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459452 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459466 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11367022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11501633 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29459539 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR1400011763.mm.1 | NR_046307.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400011763.mm.1 | NR_046307.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400011763.mm.1 | TC1400001564.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 169964_at | - | - |
| Murine Genome U74v2 Set | 169964_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 640596 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102640286 | - | - |
| MouseWG-6 v2 Expression BeadChip | 104560465 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102640286 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1253048 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102640286 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102640286 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2547989 | - | - |

4930405A10Rik
Genomic Map