Assay Details
Gene Symbol: Zfp414 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026712.3 | NP_080988.1 | 5 - 6 | 953 | 64 | |
NR_034153.1 | 6 - 7 | 1030 | 64 | |||
GenBank mRNA | AI466623.1 | - | 1 - 2 | 253 | 64 | |
AI844570.1 | - | 1 - 2 | 64 | |||
AK002706.1 | - | 5 - 6 | 943 | 64 | ||
AK169869.1 | - | 6 - 7 | 1021 | 64 | ||
BC038913.1 | - | 6 - 7 | 986 | 5099582 | 64 | |
BC116933.1 | - | 5 - 6 | 926 | 40126252 | 64 | |
BC119044.1 | - | 5 - 6 | 926 | 8733793 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31149741 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31149739 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31149742 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31149740 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P503542 | BC038913.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P503542 | NR_034153.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P503542 | AI466623.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P503542 | AI844570.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P503542 | AK002706.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P503542 | BC038913.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P503542 | NR_034153.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P503542 | AI466623.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P503542 | AI844570.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P503542 | AK002706.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 897729 | NM_175660.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 908102 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103672 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P503542 | BC038913.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P503542 | NR_034153.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P503542 | AI466623.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P503542 | AI844570.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P503542 | AK002706.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P503542 | BC038913.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P503542 | NR_034153.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P503542 | AI466623.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P503542 | AI844570.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P503542 | AK002706.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P503543 | AK002706.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P503542 | BC038913.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P503542 | NR_034153.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P503542 | AI466623.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P503542 | AI844570.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P503542 | AK002706.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P503542 | BC038913.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P503542 | NR_034153.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P503542 | AI466623.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P503542 | AI844570.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P503542 | AK002706.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149735 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149732 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149743 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149748 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149746 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149741 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149755 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149762 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149744 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149761 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149740 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149736 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149734 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149738 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149759 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149750 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149749 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149754 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149757 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149752 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31149756 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103672 | - | - |
Whole Mouse Genome G4122A | A_51_P503543 | AK002706.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P503542 | BC038913.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P503542 | NR_034153.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P503542 | AI466623.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P503542 | AI844570.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P503542 | AK002706.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P503542 | BC038913.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P503542 | NR_034153.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P503542 | AI466623.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P503542 | AI844570.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P503542 | AK002706.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103672 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13566066 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13566063 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13566068 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13105121 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13566060 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13105123 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13566069 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13566058 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31149741 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31149739 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31149742 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31149740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149825 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149821 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149816 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149813 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149819 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149807 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149811 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149817 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149788 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31149800 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31149748 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31149732 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31149761 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31149749 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31149754 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31149762 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31149743 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31149757 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31149752 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31149733 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31149744 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31149739 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31149759 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31149741 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31149756 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31149750 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31149736 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31149746 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31149755 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31149735 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10444011 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10444012 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10444014 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10444013 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10444009 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10444010 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10444015 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1436081_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10444008 | - | - |
Mouse Genome 430 2.0 Array | 1436081_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1436081_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 328801_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 328801_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1700005388.mm.1 | NM_026712.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700005388.mm.1 | NM_026712.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700005388.mm.1 | NR_034153.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700005388.mm.1 | NR_034153.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700005388.mm.1 | TC1700002775.mm.1 | MTA TC |
Mouse Expression Set 430 | 1436081_a_at | - | - |
Mouse Expression Set 430 | 1436081_PM_a_at | - | - |
Murine Genome U74v2 Set | 130439_PM_at | - | - |
Murine Genome U74v2 Set | 130439_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1436081_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1436081_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 675560 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 100670041 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1450441 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001450441 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225086 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1225086 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1259130 | - | - |
mouse-6 v1.1 (Illumina) | scl0328801.4_107-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 100670041 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1450441 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0328801.4_107-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1450441 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100670041 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1225086 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2576336 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225086 | - | - |
