Assay Details
| Gene Symbol: Gucy2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130693.3 | NP_001124165.1 | 16 - 17 | 3153 | 73 | |
| GenBank mRNA | AK131782.1 | - | 2 - 3 | 122 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gucy2d
Genomic Map