Assay Details
| Gene Symbol: Nudt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014204.1 | - | 2 - 3 | 327 | 110 | |
| BC021366.1 | - | 2 - 3 | 238 | 110 | ||
| BC056982.1 | - | 2 - 3 | 233 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK014204.1 transcripts.
Set Membership: |
|

Nudt13
Genomic Map