Assay Details
| Gene Symbol: Nudt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026341.2 | NP_080617.2 | 7 - 8 | 819 | 98 | |
| GenBank mRNA | AK014204.1 | - | 8 - 9 | 1042 | 98 | |
| BC037091.1 | - | 6 - 7 | 742 | 98 | ||
| BC040807.1 | - | 7 - 8 | 766 | 98 | ||
| BC049948.1 | - | 7 - 8 | 683 | 3666660 | 98 | |
| CA983915.1 | - | 5 - 6 | 555 | 98 | ||
| CX219422.1 | - | 2 - 3 | 227 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nudt13
Genomic Map