Assay Details
| Gene Symbol: Ppp1r11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029632.3 | NP_083908.1 | 2 - 3 | 481 | 72 | |
| GenBank mRNA | AK005379.1 | - | 2 - 3 | 378 | 72 | |
| AK077455.1 | - | 2 - 3 | 364 | 72 | ||
| AK089107.1 | - | 2 - 3 | 364 | 72 | ||
| AK162607.1 | - | 2 - 3 | 365 | 72 | ||
| AK170104.1 | - | 2 - 3 | 364 | 72 | ||
| BC027737.1 | - | 2 - 3 | 351 | 5003418 | 72 | |
| EF528576.1 | - | 2 - 3 | 195 | 72 | ||
| EF528578.1 | - | 2 - 3 | 180 | 72 | ||
| EF528579.1 | - | 1 - 2 | 93 | 72 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166376 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166413 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166379 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166409 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166375 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166412 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166410 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166377 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166378 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166374 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166408 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P110791 | AK162607.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P110791 | AK170104.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P110791 | BC027737.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P110791 | NM_029632.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P110791 | AK005379.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 947465 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 755299 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 214902 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 117078 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114499 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P110791 | AK162607.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P110791 | AK170104.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P110791 | BC027737.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P110791 | NM_029632.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P110791 | AK005379.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166440 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166438 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166452 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166442 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166447 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166432 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114499 | - | - |
| Whole Mouse Genome G4122A | A_51_P105320 | AK162607.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P105320 | BB842861.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P105320 | NM_029632.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P105320 | AK005379.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P105320 | AK077455.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110791 | AK162607.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110791 | AK170104.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110791 | BC027737.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110791 | NM_029632.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110791 | AK005379.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114499 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13447233 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13447239 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13447249 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13447247 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13447251 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166412 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166379 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166377 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166375 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166408 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166410 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166378 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166376 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166374 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166409 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166391 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166426 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166423 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166437 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166455 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13447233 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166448 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166432 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166457 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166442 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166435 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166436 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13277684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166450 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13277703 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10450816 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450818 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450817 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450815 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448565_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417060_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10450814 | - | - |
| Mouse Genome 430 2.0 Array | 1448565_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448565_at | - | - |
| Mouse Genome 430 2.0 Array | 1417060_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417060_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76497_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76497_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700019448.mm.1 | NM_029632.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019448.mm.1 | NM_029632.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019448.mm.1 | TC1700002041.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700019446.mm.1 | NM_029632.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019446.mm.1 | NM_029632.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019446.mm.1 | TC1700002041.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700019447.mm.1 | NM_029632.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019447.mm.1 | NM_029632.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019447.mm.1 | TC1700002041.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448565_PM_at | - | - |
| Mouse Expression Set 430 | 1417060_PM_at | - | - |
| Mouse Expression Set 430 | 1417060_at | - | - |
| Mouse Expression Set 430 | 1448565_at | - | - |
| Murine Genome U74v2 Set | 166497_at | - | - |
| Murine Genome U74v2 Set | 166497_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448565_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417060_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448565_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417060_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417060_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448565_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448565_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417060_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100380113 | - | - |
| MouseWG-6 v2 Expression BeadChip | 3610072 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 100380113 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3610072 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003610072 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2757489 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2757489 | - | - |
| mouse-6 v1.1 (Illumina) | scl0076497.2_72-S | - | - |
| mouse-6 v1.1 (Illumina) | scl49943.4_9-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 100380113 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3610072 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0076497.2_72-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3610072 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100380113 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2757489 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2757489 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2816137 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0076497.2_72-S | - | - |

Ppp1r11
Genomic Map