Assay Details
| Gene Symbol: Mrps18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026768.3 | NP_081044.1 | 4 - 5 | 388 | 66 | |
| GenBank mRNA | AB049953.1 | - | 4 - 5 | 397 | 66 | |
| AK003444.1 | - | 4 - 5 | 389 | 66 | ||
| AK028226.1 | - | 4 - 5 | 390 | 66 | ||
| AK172171.1 | - | 4 - 5 | 383 | 66 | ||
| BC025456.1 | - | 2 - 3 | 184 | 66 | ||
| BC041151.1 | - | 1 - 2 | 137 | 66 | ||
| BC058959.1 | - | 4 - 5 | 376 | 6817698 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrps18a
Genomic Map