Assay Details
Gene Symbol: Klc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029091.2 | NP_083367.1 | 9 - 10 | 1320 | 66 | |
GenBank mRNA | AK004774.1 | - | 9 - 10 | 1321 | 66 | |
AK170793.1 | - | 9 - 10 | 1304 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
