Assay Details
Gene Symbol: Klc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029091.2 | NP_083367.1 | 10 - 11 | 1379 | 86 | |
GenBank mRNA | AK004774.1 | - | 10 - 11 | 1380 | 86 | |
AK170793.1 | - | 10 - 11 | 1363 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
