Assay Details
| Gene Symbol: Atat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142744.1 | NP_001136216.1 | 9 - 10 | 758 | 134 | |
| NM_001142745.1 | NP_001136217.1 | 9 - 10 | 758 | 134 | ||
| NM_028476.4 | NP_082752.3 | 8 - 9 | 689 | 134 | ||
| GenBank mRNA | AK011750.2 | - | 9 - 10 | 734 | 134 | |
| AK011838.1 | - | 9 - 10 | 747 | 134 | ||
| AK014265.1 | - | 9 - 10 | 732 | 134 | ||
| AK028297.1 | - | 9 - 10 | 748 | 134 | ||
| AK034725.1 | - | 8 - 9 | 667 | 134 | ||
| AK042804.1 | - | 3 - 4 | 184 | 134 | ||
| AK049682.1 | - | 8 - 9 | 639 | 134 | ||
| AK133694.1 | - | 9 - 10 | 735 | 134 | ||
| BC028847.1 | - | 9 - 10 | 737 | 4007862 | 134 | |
| BC030669.1 | - | 8 - 9 | 667 | 1246856 | 134 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Atat1
Genomic Map