Assay Details
| Gene Symbol: Daam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008231.2 | NP_001008232.2 | 18 - 19 | 2484 | 106 | |
| GenBank mRNA | AK009382.1 | - | 3 - 4 | 305 | 106 | |
| AK122266.1 | - | 16 - 17 | 5402 | 106 | ||
| AY426536.1 | - | 18 - 19 | 2513 | 106 | ||
| BC037720.1 | - | 2 - 3 | 205 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Daam2
Genomic Map