Assay Details
| Gene Symbol: Dhx16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026987.2 | NP_081263.2 | 14 - 15 | 2452 | 63 | |
| GenBank mRNA | AK010431.1 | - | 1 - 2 | 70 | 63 | |
| AK122314.2 | - | 14 - 15 | 2387 | 63 | ||
| BC009147.1 | - | 14 - 15 | 2411 | 2647184 | 63 | |
| BC046781.1 | - | 14 - 15 | 2380 | 5716182 | 63 | |
| BC138204.1 | - | 14 - 15 | 2391 | 8861223 | 63 | |
| BC138205.1 | - | 14 - 15 | 2391 | 8861224 | 63 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dhx16
Genomic Map