Assay Details
| Gene Symbol: Metap1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025633.3 | NP_079909.1 | 8 - 9 | 877 | 80 | |
| GenBank mRNA | AJ414378.1 | - | 5 - 6 | 740 | 80 | |
| AK005241.1 | - | 8 - 9 | 874 | 80 | ||
| AK010067.1 | - | 8 - 9 | 877 | 80 | ||
| BC051534.1 | - | 5 - 6 | 751 | 3466745 | 80 | |
| BC060739.1 | - | 7 - 8 | 2385 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Metap1d
Genomic Map