Assay Details
| Gene Symbol: Hpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017371.2 | NP_059067.2 | 1 - 2 | 106 | 67 | |
| GenBank mRNA | AK145928.1 | - | 1 - 2 | 104 | 67 | |
| BY088352.1 | - | 1 - 2 | 107 | 67 | ||
| U89889.1 | - | 1 - 2 | 62 | 67 | ||
More Information
Set Membership: |
|

Hpx
Genomic Map