Assay Details
| Gene Symbol: Olfr560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147113.2 | NP_667324.2 | 1 | 461 | 132 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046463 | NM_147113.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046463 | NM_147113.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P216165 | NM_147113.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380512 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380515 | - | - |
| Whole Mouse Genome G4122A | A_51_P216165 | NM_147113.2 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06406048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06599504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06228489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05952650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05891396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25380501 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10566179 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10566180 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10566172 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700032669.mm.1 | NM_147113.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032669.mm.1 | NM_147113.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032669.mm.1 | TC0700003869.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 670315 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000670315 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2653364 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2653364 | - | - |
| mouse-6 v1.1 (Illumina) | scl0259117.1_37-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 670315 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0259117.1_37-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 670315 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1240724 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1240724 | - | - |

Olfr560
Genomic Map