Assay Details
Gene Symbol: Atg16l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111111.1 | NP_001104581.1 | 7 - 8 | 894 | 95 | |
GenBank mRNA | AB476648.1 | - | 7 - 8 | 833 | 95 | |
AK155125.1 | - | 7 - 8 | 884 | 95 | ||
AK157410.1 | - | 7 - 8 | 873 | 95 | ||
AK158186.1 | - | 7 - 8 | 834 | 95 | ||
BC060054.1 | - | 7 - 8 | 1057 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
