Assay Details
| Gene Symbol: Atg16l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111111.1 | NP_001104581.1 | 8 - 9 | 954 | 62 | |
| GenBank mRNA | AB476648.1 | - | 8 - 9 | 893 | 62 | |
| AK155125.1 | - | 8 - 9 | 944 | 62 | ||
| AK157410.1 | - | 8 - 9 | 933 | 62 | ||
| AK158186.1 | - | 8 - 9 | 894 | 62 | ||
| BC060054.1 | - | 8 - 9 | 1117 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atg16l2
Genomic Map