Assay Details
| Gene Symbol: Atg16l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111111.1 | NP_001104581.1 | 11 - 12 | 1246 | 63 | |
| GenBank mRNA | AB476647.1 | - | 10 - 11 | 1122 | 63 | |
| AB476648.1 | - | 11 - 12 | 1185 | 63 | ||
| AK010773.1 | - | 10 - 11 | 1167 | 63 | ||
| AK131109.1 | - | 1016 | 63 | |||
| AK155125.1 | - | 11 - 12 | 1236 | 63 | ||
| AK157410.1 | - | 11 - 12 | 1225 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atg16l2
Genomic Map