Assay Details
Gene Symbol: Plau | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008873.3 | NP_032899.1 | 3 - 4 | 196 | 67 | |
GenBank mRNA | AK054349.1 | - | 3 - 4 | 195 | 67 | |
BC120709.1 | - | 2 - 3 | 136 | 40129632 | 67 | |
BC120713.1 | - | 2 - 3 | 136 | 40129636 | 67 | |
X02389.1 | - | 3 - 4 | 151 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
