Assay Details
Gene Symbol: Plau | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008873.3 | NP_032899.1 | 6 - 7 | 556 | 75 | |
GenBank mRNA | AK054349.1 | - | 6 - 7 | 555 | 75 | |
BC120709.1 | - | 5 - 6 | 496 | 40129632 | 75 | |
BC120713.1 | - | 5 - 6 | 496 | 40129636 | 75 | |
BC145037.1 | - | 5 - 6 | 519 | 75 | ||
X02389.1 | - | 6 - 7 | 511 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
