Assay Details
| Gene Symbol: Eif3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025344.2 | NP_079620.2 | 2 - 3 | 465 | 108 | |
| GenBank mRNA | AK002778.1 | - | 2 - 3 | 464 | 108 | |
| AK003372.1 | - | 2 - 3 | 463 | 108 | ||
| AK010333.1 | - | 2 - 3 | 459 | 108 | ||
| AK088537.1 | - | 2 - 3 | 465 | 108 | ||
| AK136797.1 | - | 464 | 108 | |||
| AK152037.1 | - | 2 - 3 | 466 | 108 | ||
| AK190701.1 | - | 2 - 3 | 110 | 108 | ||
| AK197956.1 | - | 2 - 3 | 112 | 108 | ||
| AK217928.1 | - | 1 - 2 | 83 | 108 | ||
| BC070473.1 | - | 2 - 3 | 447 | 30634857 | 108 | |
| BC083190.1 | - | 2 - 3 | 450 | 30358339 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eif3f
Genomic Map