Assay Details
| Gene Symbol: Olfr609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147082.2 | NP_667293.2 | 1 | 818 | 77 | |
| GenBank mRNA | BC128008.1 | - | 1 | 822 | 40135990 | 77 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281608 | NM_147082.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P281608 | NM_147082.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141008 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281608 | NM_147082.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P281608 | NM_147082.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P281608 | NM_147082.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P281608 | NM_147082.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383512 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383509 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25383543 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141008 | - | - |
| Whole Mouse Genome G4122A | A_51_P281608 | NM_147082.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P281608 | NM_147082.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107599 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141008 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06546213 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06546194 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06546202 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06546193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25383554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383543 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06030017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383512 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06546214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383540 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06546211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05946746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06546210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06546202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25383507 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10566218 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10566217 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700032716.mm.1 | NM_147082.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032716.mm.1 | NM_147082.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700032716.mm.1 | TC0700003892.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 352669 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2802733 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2802733 | - | - |

Olfr609
Genomic Map