Assay Details
| Gene Symbol: Far1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285831.1 | NP_001272760.1 | 9 - 10 | 1116 | 94 | |
| GenBank mRNA | AK033674.1 | - | 9 - 10 | 1059 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Far1
Genomic Map