Assay Details
| Gene Symbol: Guca1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146079.1 | NP_666191.1 | 1 - 2 | 307 | 61 | |
| GenBank mRNA | AK033052.1 | - | 1 - 2 | 272 | 61 | |
| BC018258.1 | - | 1 - 2 | 235 | 4511650 | 61 | |
| BC018480.1 | - | 1 - 2 | 276 | 4500919 | 61 | |
| BC029017.1 | - | 1 - 2 | 252 | 5359531 | 61 | |
| BC029661.1 | - | 1 - 2 | 262 | 5366068 | 61 | |
| BC031367.1 | - | 1 - 2 | 270 | 5358667 | 61 | |
| BC031916.1 | - | 1 - 2 | 307 | 5369642 | 61 | |
| BC033348.1 | - | 1 - 2 | 257 | 61 | ||
| BC033358.1 | - | 1 - 2 | 306 | 5368968 | 61 | |
| BC033364.1 | - | 1 - 2 | 257 | 61 | ||
| BC033380.1 | - | 1 - 2 | 262 | 5366953 | 61 | |
| BC033383.1 | - | 1 - 2 | 263 | 5368064 | 61 | |
| BC033396.1 | - | 1 - 2 | 254 | 61 | ||
| BC033423.1 | - | 1 - 2 | 259 | 61 | ||
| BC034331.1 | - | 1 - 2 | 263 | 5357797 | 61 | |
| BC034595.1 | - | 1 - 2 | 262 | 5362191 | 61 | |
| Gene Symbol: Actr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC027134.1 | - | 226 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Guca1b
Genomic Map