Assay Details
| Gene Symbol: Prkcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008855.2 | NP_032881.1 | 3 - 4 | 512 | 75 | |
| GenBank mRNA | BC127083.1 | - | 3 - 4 | 331 | 40111744 | 75 |
| CJ050078.1 | - | 3 - 4 | 303 | 75 | ||
| X53532.1 | - | 3 - 4 | 340 | 75 | ||
| X59274.1 | - | 3 - 4 | 328 | 75 | ||
More Information
Set Membership: |
|

Prkcb
Genomic Map