Assay Details
| Gene Symbol: Gnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008136.2 | NP_032162.2 | 4 - 5 | 745 | 98 | |
| GenBank mRNA | BC010298.1 | - | 4 - 5 | 700 | 3155411 | 98 |
| BC013243.1 | - | 4 - 5 | 769 | 98 | ||
| BC094391.1 | - | 2 - 3 | 182 | 98 | ||
| BC129969.1 | - | 2 - 3 | 277 | 98 | ||
| BC129970.1 | - | 2 - 3 | 277 | 98 | ||
| BM949933.1 | - | 4 - 5 | 491 | 98 | ||
| X65026.1 | - | 3 - 4 | 388 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the X65026.1 transcripts.
Set Membership: |
|

Gnl1
Genomic Map