Assay Details
| Gene Symbol: Gnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008136.2 | NP_032162.2 | 7 - 8 | 1123 | 77 | |
| GenBank mRNA | AK078187.1 | - | 7 - 8 | 1086 | 77 | |
| BC010298.1 | - | 7 - 8 | 1078 | 3155411 | 77 | |
| BC013243.1 | - | 7 - 8 | 1147 | 77 | ||
| BC094391.1 | - | 5 - 6 | 560 | 77 | ||
| BC110397.1 | - | 3 - 4 | 388 | 77 | ||
| BC129969.1 | - | 5 - 6 | 655 | 77 | ||
| BC129970.1 | - | 5 - 6 | 655 | 77 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gnl1
Genomic Map