Assay Details
| Gene Symbol: Bcl7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009746.2 | NP_033876.1 | 1 - 2 | 332 | 76 | |
| GenBank mRNA | AK049064.1 | - | 1 - 2 | 352 | 76 | |
| AK131935.1 | - | 1 - 2 | 221 | 76 | ||
| BC005673.1 | - | 1 - 2 | 332 | 3153237 | 76 | |
| BC058530.1 | - | 1 - 2 | 203 | 6818556 | 76 | |
| Y11905.1 | - | 2 - 3 | 150 | 76 | ||
More Information
Set Membership: |
|

Bcl7c
Genomic Map