Assay Details
Gene Symbol: Atp2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007504.2 | NP_031530.2 | 19 - 20 | 2893 | 74 | |
GenBank mRNA | AK132537.1 | - | 4 - 5 | 610 | 74 | |
AK160497.1 | - | 2 - 3 | 182 | 74 | ||
AY081946.1 | - | 19 - 20 | 2924 | 74 | ||
BC036292.1 | - | 19 - 20 | 2893 | 4191741 | 74 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
