Assay Details
| Gene Symbol: Setd1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178029.3 | NP_821172.2 | 16 - 17 | 4856 | 65 | |
| GenBank mRNA | BC010250.1 | - | 3 - 4 | 518 | 65 | |
| BC042890.1 | - | 3 - 4 | 944 | 65 | ||
| BC049883.1 | - | 9 - 10 | 2244 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Setd1a
Genomic Map