Assay Details
| Gene Symbol: Nfe2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK029360.1 | - | 1 - 2 | 153 | 61 | |
| BC057948.1 | - | 1 - 2 | 124 | 61 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nfe2l2| Gene Symbol: Nfe2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK029360.1 | - | 1 - 2 | 153 | 61 | |
| BC057948.1 | - | 1 - 2 | 124 | 61 | ||
Set Membership: |
|